Palladium ICP Standard, Inorganic Ventures
Supplier: INORGANIC VENTURES MS
CGPDN1-500ML CGPD1-500ML
100360-634EA 1185.69 USD
100360-634 100360-632
Palladium ICP Standard, Inorganic Ventures
Standards Cationic Standards Palladium Standards
Certified reference material for Palladium, traceable to NIST.
One year expiration date (defined by supplier as the amount of time that it should remain in use after opening).