Boron ICP Standard, Inorganic Ventures
Supplier: INORGANIC VENTURES MS
CGB10-500ML CGB1-500ML
100359-904EA 208.92 USD
100359-904 100360-584
Boron ICP Standard, Inorganic Ventures
Standards Cationic Standards Boron Standards
Certified reference material for Boron, traceable to NIST.
One year expiration date (defined by supplier as the amount of time that it should remain in use after opening).