Brucella Agar/Broth, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
D04
J87
10068-016PK
389.62
USD
10068-016
10324-448
Brucella Agar/Broth, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
For the cultivation of anaerobic bacteria and the selective isolation of Bacteroides spp.
- Broth for preservation of organisms by freezing
- Cultivation of anaerobic bacteria
- Selective isolation of Bacteroides
For the cultivation of anaerobic bacteria and the selective isolation of Bacteroides spp. Brucella Broth with 15% Glycerol for the preservation of organisms by freezing, packaged in a glass cryovial with a black screw-cap.