Pacific Blue™ Annexin V Apoptosis Detection Kit, Biolegend®
Supplier: Biolegend
640922 640928
10763-246EA 368.9 USD
10763-246 10763-252
Pacific Blue™ Annexin V Apoptosis Detection Kit, Biolegend®
Assays Cellular Assays
Annexin V Apoptosis detection kit has been specifically designed for the identification of apoptotic and necrotic cells.
  • Binding Buffer is recommended for use with Annexin V staining
  • Target and identify apoptotic cells
  • Intracellular proteins that binds to phosphatidylserine (PS) in a calcium-dependent manner
Annexin V (or Annexin A5) is a member of the annexin family of intracellular proteins that binds to phosphatidylserine (PS) in a calcium-dependent manner
Annexin V Binding Buffer is recommended for use with Annexin V staining. Annexin V binding alone cannot differentiate between apoptotic and necrotic cells. To help distinguish between the necrotic and apoptotic cells we recommend use of our Propidium Iodide Solution (PI). Early apoptotic cells will exclude PI, while late stage apoptotic cells and necrotic cells will stain positively, due to the passage of these dyes into the nucleus where they bind to DNA.
Propidium iodide is a fluorescent dye that binds to DNA. When excited by 488 nm laser light, it can be detected with in the PE/Texas Red® channel with a bandpass filter 610/10. It is commonly used in evaluation of cell viability or DNA content in cell cycle analysis by flow cytometry. 7-AAD (7-amino-actinomycin D) has a high DNA binding constant and is efficiently excluded by intact cells. It is useful for DNA analysis and dead cell discrimination during flow cytometric analysis. When excited by 488 laser light, 7-AAD fluorescence is detected in the far red range of the spectrum (650 nm long-pass filter).
PS is normally only found on the intracellular leaflet of the plasma membrane in healthy cells, but during early apoptosis, membrane asymmetry is lost and PS translocates to the external leaflet. Fluorochrome-labeled Annexin V can then be used to specifically target and identify apoptotic cells.