Standard Series™ NMR Tubes, 5 mm, NORELL
Supplier: Norell Inc
502-8 552-7 509-UP-8 506-P-8 505-P-7 ST500-8 ST550-7 508-UP-7 ST550-8 507-HP-8 552-8 507-HP-7 506-P-7 ST500-7 509-UP-7 505-P-8
100554-266PK 261.52 USD
100554-266 101190-186 100554-244 100554-254 100554-260 100554-270 101190-190 100554-246 101190-192 100554-176 101190-188 100554-250 100554-252 100554-268 100554-242 100554-262
Standard Series™ NMR Tubes, 5 mm, NORELL
Tubes NMR Tubes
Manufactured from ASTM Type 1 Class B glass.
  • Designed for routine NMR
  • Tube bottoms are uniformly hemispherical and consistent
  • Minimizes shimming and susceptibility differences among samples