Comply™ Chemical Integrators, 3M™
Supplier: Healthcare Products
MMM 1243B MMM 1243RE MMM 41360
75786-420CS 390.42 USD
75786-420 75787-962 75787-964
Comply™ Chemical Integrators, 3M™
Sterilization Indicators
Comply™ Chemical Integrators provides an immediate, accurate, and easy-to-read method of monitoring sterilization process conditions inside each pack.
It can be used in all steam sterilizers. The easy-to-read moving front style has been shown to have a higher reading accuracy than color interpretation style indicators.