Specimen Clamps and Accessories, Thermo Scientific
Supplier: Epredia
715040 715070 715280 715320 715370 716050 715380
16002-440EA 1095.97 USD
16002-440 16002-442 16002-462 16002-472 16002-444 16002-446 16002-470 16002-474 16002-508 16002-476 16002-448
Specimen Clamps and Accessories, Thermo Scientific
Specimen Mounting Supplies
Get the most out of your Thermo Scientific™ microtome with this Thermo Scientific™ Specimen Clamp and Accessories.
  • Works in addition to specimen orientation in the X- and Y-axis
  • Settings via drive mechanism
  • Tandem spring-loaded system ensures easy insertion of the specimen without tilting or squeezing
  • Easy removal of the round specimen
  • When working with round specimen clamps, the standard specimen clamp must be equipped with a jaw light
  • Spring-loaded jaws secure the specimen in position
Segment Arc for universal specimen holder with 360° rotation and standard accurate orientation of 30°.
Universal Specimen Holder for flat and round specimens up to 8mm used with the segment arc.
Standard Specimen Clamp with fixed upper jaw ensuring greatest stability, orientation on X- and Y-axes, and rotation for accurate positioning.
Insert for Round Specimens to be fitted in the standard specimen clamp.
V-Distance Piece for standard specimen clamps.
Specimen Stages work with standard specimen clamp with V-insert.
Universal Cassette Clamp with clamping jaws to be inserted in the microtome standard specimen clamp.
Foil Clamp to be fitted in the standard specimen clamp.