LymphoSep™ Lymphocyte Separation Medium, MP Biomedicals
Supplier: MP Biomedicals
091692249
10220-620EA 61.99 USD
10220-620
LymphoSep™ Lymphocyte Separation Medium, MP Biomedicals
Cell Separation Media
LymphoSep™ - Lymphocyte separation medium is a sterile, iso-osmotic, low viscosity solution for density centrifugation yielding a clear separation of lymphocytes from heparinized or defibrinated human blood.
  • In vitro isolation of lymphocytes from diluted whole blood
  • Maximum yield of mononuclear cells
  • Easy one-step centrifugation
  • >96% cell viability of lymphocytes
  • Low endotoxins
  • Autoclave sterilization
Lymphocytes are isolated by layering diluted, heparinized blood over a solution of sodium metrizoate that contains a polysaccharide. This polysaccharide aggregates the erythrocytes so that they can be sedimented by low centrifugation. The less dense lymphocytes remain as a clearly defined band at the plasma LymphoSep interface.
Precaución: Store at room temperature (15...30 °C), protect from light.