1-(5-Bromo-7-azaindole)ethanone ≥95%
Supplier: Matrix Scientific
Synonyms:
1-(5-Bromo-1H-pyrrolo[2,3-b]pyridin-3-yl)ethanone
116010-1G
116010-10G
102900-180EA
989.23
USD
102900-180
102900-184
1-(5-Bromo-7-azaindole)ethanone ≥95%
1-(5-Bromo-7-azaindole)ethanone
Hazard: Irritant
Formula:
C₉H₇BrN₂O MW: 239.08 g/mol |
MDL Number:
MFCD12403936 CAS Number: 866545-96-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...