5-Boc-2-oxa-5-azaspiro[3.5]nonane ≥95%

Supplier: Matrix Scientific

115844-250MG 115844-500MG 115844-1G
102898-510EA 1098.46 USD
102898-510 102898-512 102898-514
5-Boc-2-oxa-5-azaspiro[3.5]nonane ≥95%
5-Boc-2-oxa-5-azaspiro[3.5]nonane

Hazard: Irritant

Formula: C₁₂H₂₁NO₃
MW: 227.31 g/mol
MDL Number: MFCD22628722
CAS Number: 1374659-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR