5-Boc-2,8-dioxa-5-azaspiro[3.5]nonane ≥95%

Supplier: Matrix Scientific

115842-500MG 115842-1G 115842-250MG
102898-326EA 3489.23 USD
102898-326 102898-328 102899-764
5-Boc-2,8-dioxa-5-azaspiro[3.5]nonane ≥95%
5-Boc-2,8-dioxa-5-azaspiro[3.5]nonane

Hazard: Irritant

Formula: C₁₁H₁₉NO₄
MW: 229.28 g/mol
MDL Number: MFCD22121388
CAS Number: 1374658-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR