3-Oxa-9-azabicyclo[3.3.1]nonane-7-carboxylic acid methyl ester hydrochloride ≥95%

Supplier: Matrix Scientific

115795-250MG 115795-500MG 115795-1G
102899-678EA 1992.31 USD
102899-678 102899-680 102899-682
3-Oxa-9-azabicyclo[3.3.1]nonane-7-carboxylic acid methyl ester hydrochloride ≥95%
3-Oxa-9-azabicyclo[3.3.1]nonane-7-carboxylic acid methyl ester hydrochloride

Hazard: Irritant

Formula: C₉H₁₆ClNO₃
MDL Number: MFCD19707045
CAS Number: 1524708-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR