4,4-Dimethyl-2-oxopentanoic acid ethyl ester ≥95%

Supplier: Matrix Scientific

115773-500MG 115773-1G 115773-5G
102899-658EA 578.46 USD
102899-658 102899-660 102899-662
4,4-Dimethyl-2-oxopentanoic acid ethyl ester ≥95%
4,4-Dimethyl-2-oxopentanoic acid ethyl ester

Hazard: Irritant

Formula: C₉H₁₆O₃
MDL Number: MFCD22573384
CAS Number: 929879-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR