Methyl-4-amino-1H-pyrazole-3-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 4-amino-1H-pyrazole-3-carboxylate
115721-1G
115721-5G
102899-478EA
798.46
USD
102899-478
102899-480
Methyl-4-amino-1H-pyrazole-3-carboxylate ≥95%
Methyl-4-amino-1H-pyrazole-3-carboxylate
Hazard: Irritant
Formula:
C₅H₇N₃O₂ MW: 141.13 g/mol |
MDL Number:
MFCD02646737 CAS Number: 360056-45-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...