Methyl-6-bromo-1H-imidazo[1,2-a]pyridine-8-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 6-bromoimidazo[1,2-a]pyridine-8-carboxylate
115643-1G
115643-5G
102897-044EA
989.23
USD
102897-044
102897-046
Methyl-6-bromo-1H-imidazo[1,2-a]pyridine-8-carboxylate ≥95%
Methyl-6-bromo-1H-imidazo[1,2-a]pyridine-8-carboxylate
Hazard: Irritant
Formula:
C₉H₇BrN₂O₂ MW: 255.08 g/mol |
MDL Number:
MFCD13195317 CAS Number: 908581-18-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...