tert-Butyl-3-fluoro-4-((hydroxyimino)methyl)-4-ethylpiperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

115633-250MG 115633-500MG 115633-1G
102899-514EA 4064.62 USD
102899-514 102899-516 102899-518
tert-Butyl-3-fluoro-4-((hydroxyimino)methyl)-4-ethylpiperidine-1-carboxylate ≥95%
tert-Butyl-3-fluoro-4-((hydroxyimino)methyl)-4-ethylpiperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₂H₂₁FN₂O₃
MW: 260.31 g/mol
MDL Number: MFCD22421712
CAS Number: 1400765-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR