S-(3,3-Difluorocyclobutyl)ethanethioic acid ester ≥95%

Supplier: Matrix Scientific

115579-100MG 115579-250MG 115579-500MG
102896-878EA 769.23 USD
102896-878 102896-880 102896-882
S-(3,3-Difluorocyclobutyl)ethanethioic acid ester ≥95%
S-(3,3-Difluorocyclobutyl)ethanethioic acid ester

Hazard: Irritant

Formula: C₆H₈F₂OS
MDL Number: MFCD22421697
CAS Number: 1310729-92-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR