2-Amino-6-azaspiro[3.4]octane-6-carboxylic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

115578-500MG 115578-1G
102896-874EA 3515.38 USD
102896-874 102896-876
2-Amino-6-azaspiro[3.4]octane-6-carboxylic acid tert-butyl ester ≥95%
2-Amino-6-azaspiro[3.4]octane-6-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₂H₂₂N₂O₂
MDL Number: MFCD17016203
CAS Number: 1239319-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR