7-Bromo[1,2,4]triazolo[1,5-a]pyridin-2-amine ≥95%
Supplier: Matrix Scientific
Synonyms:
7-Bromo-[1,2,4]triazolo[1,5-a]pyridin-2-amine
115574-1G
115574-5G
115574-25G
102899-466EA
495.38
USD
102899-466
102899-468
102899-494
7-Bromo[1,2,4]triazolo[1,5-a]pyridin-2-amine ≥95%
7-Bromo[1,2,4]triazolo[1,5-a]pyridin-2-amine
Hazard: Irritant
Formula:
C₆H₅BrN₄ MW: 213.0347 g/mol |
MDL Number:
MFCD11846606 CAS Number: 882521-63-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...