Allyl-4-methylbenzenesulfonate ≥95%

Supplier: Matrix Scientific

Synonyms: Allyl 4-methylbenzenesulphonate

115573-1G 115573-5G
102899-462EA 220 USD
102899-462 102899-464
Allyl-4-methylbenzenesulfonate ≥95%
Allyl-4-methylbenzenesulfonate

Hazard: Irritant

Formula: C₁₀H₁₂O₃S
MW: 212.27 g/mol
MDL Number: MFCD00093777
CAS Number: 4873-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR