5-(Tributylstannyl)-2-pyridinecarbonitrile ≥95%

Supplier: Matrix Scientific

115572-1G 115572-5G 115572-25G
102899-456EA 330.77 USD
102899-456 102899-458 102899-460
5-(Tributylstannyl)-2-pyridinecarbonitrile ≥95%
5-(Tributylstannyl)-2-pyridinecarbonitrile

Hazard: Irritant

Formula: C₁₈H₃₀N₂Sn
MDL Number: MFCD22421738
CAS Number: 866606-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR