3-Fluoro-4-(tributylstannyl)pyridine ≥95%
Supplier: Matrix Scientific
115559-1G
115559-5G
115559-25G
102896-698EA
64.62
USD
102896-698
102896-700
102896-702
3-Fluoro-4-(tributylstannyl)pyridine ≥95%
3-Fluoro-4-(tributylstannyl)pyridine
Hazard: Irritant
Formula:
C₁₇H₃₀FNSn |
CAS Number:
259807-88-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...