Azetidin-3-yl(pyrrolidin-1-yl)methanone ≥98%

Supplier: Matrix Scientific

115513-100MG 115513-250MG
102896-576EA 1318.46 USD
102896-576 102896-578
Azetidin-3-yl(pyrrolidin-1-yl)methanone ≥98%
Azetidin-3-yl(pyrrolidin-1-yl)methanone

Hazard: Irritant

Formula: C₈H₁₄N₂O
MDL Number: MFCD21650437
CAS Number: 1257293-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR