2-Azabicyclo[2.2.2]octane-2,6-dicarboxylic acid-2-tert-butyl ester ≥95%

Supplier: Matrix Scientific

115512-250MG 115512-500MG 115512-1G
102896-570EA 1647.69 USD
102896-570 102896-572 102896-574
2-Azabicyclo[2.2.2]octane-2,6-dicarboxylic acid-2-tert-butyl ester ≥95%
2-Azabicyclo[2.2.2]octane-2,6-dicarboxylic acid-2-tert-butyl ester

Hazard: Irritant

Formula: C₁₃H₂₁NO₄
MW: 255.32 g/mol
MDL Number: MFCD18089871
CAS Number: 1250997-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR