N,N-Diethyl-N-methyl-N-(2-methoxyethyl)ammonium imidodisulfuryl fluoride ≥98%
Supplier: Matrix Scientific
Synonyms:
Diethyl(2-methoxyethyl)methylammonium Bis(fluorosulphonyl)imide
115476-1G
115476-5G
102896-440EA
330.77
USD
102896-440
102896-442
N,N-Diethyl-N-methyl-N-(2-methoxyethyl)ammonium imidodisulfuryl fluoride ≥98%
N,N-Diethyl-N-methyl-N-(2-methoxyethyl)ammonium imidodisulfuryl fluoride
Hazard: Irritant
Formula:
C₈H₂₀F₂N₂O₅S₂ MW: 326.38 g/mol |
MDL Number:
MFCD22421701 CAS Number: 1079129-48-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...