4-(Benzyloxy)-2,6-dimethylbenzaldehyde ≥97%

Supplier: Matrix Scientific

115229-500MG 115229-1G
102895-646EA 624.62 USD
102895-646 102895-648
4-(Benzyloxy)-2,6-dimethylbenzaldehyde ≥97%
4-(Benzyloxy)-2,6-dimethylbenzaldehyde

Hazard: Irritant

Formula: C₁₆H₁₆O₂
Melting Pt: 60…62 °C
MDL Number: MFCD06797237
CAS Number: 28924-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR