8-(4-Aminopiperidin-1-yl)[1,2,4]triazolo[4,3-a]pyrazin-3(2H)-one ≥95%
Supplier: Matrix Scientific
115179-500MG
115179-1G
102895-526EA
693.85
USD
102895-526
102895-528
8-(4-Aminopiperidin-1-yl)[1,2,4]triazolo[4,3-a]pyrazin-3(2H)-one ≥95%
8-(4-Aminopiperidin-1-yl)[1,2,4]triazolo[4,3-a]pyrazin-3(2H)-one
Hazard: Irritant
Formula:
C₁₀H₁₄N₆O MW: 234.26 g/mol |
MDL Number:
MFCD18915349 CAS Number: 1325305-68-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...