1-[6-(3-Fluorophenyl)pyridazin-3-yl]piperidin-4-amine ≥95%

Supplier: Matrix Scientific

115138-500MG 115138-1G
102895-476EA 693.85 USD
102895-476 102895-478
1-[6-(3-Fluorophenyl)pyridazin-3-yl]piperidin-4-amine ≥95%
1-[6-(3-Fluorophenyl)pyridazin-3-yl]piperidin-4-amine

Hazard: Irritant

Formula: C₁₅H₁₇FN₄
MDL Number: MFCD25951460
CAS Number: 1338674-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR