5-(4-Fluorophenyl)-2,2-dimethyl-2,3-dihydro-4H-imidazol-4-one 97
Supplier: Matrix Scientific
115091-500MG
115091-1G
102891-578EA
693.85
USD
102891-578
102891-580
5-(4-Fluorophenyl)-2,2-dimethyl-2,3-dihydro-4H-imidazol-4-one 97
5-(4-Fluorophenyl)-2,2-dimethyl-2,3-dihydro-4H-imidazol-4-one
Hazard: Irritant
Formula:
C₁₁H₁₁FN₂O MW: 206.22 g/mol |
MDL Number:
MFCD19104837 CAS Number: 899926-79-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...