Ethyl-3-{[4-(4-chlorophenyl)piperazin-1-yl]sulfonyl}-1H-pyrazole-4-carboxylate ≥90%

Supplier: Matrix Scientific

115072-500MG 115072-1G
102895-358EA 693.85 USD
102895-358 102895-360
Ethyl-3-{[4-(4-chlorophenyl)piperazin-1-yl]sulfonyl}-1H-pyrazole-4-carboxylate ≥90%
Ethyl-3-{[4-(4-chlorophenyl)piperazin-1-yl]sulfonyl}-1H-pyrazole-4-carboxylate

Hazard: Irritant

Formula: C₁₆H₁₉ClN₄O₄S
MW: 398.87 g/mol
MDL Number: MFCD18194700
CAS Number: 1261016-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR