tert-Butyl-2-phenyl-3-thioxo-1,4,8-triazaspiro[4.5]dec-1-ene-8-carboxylate ≥97%

Supplier: Matrix Scientific

115054-1G 115054-500MG
102891-270EA 1040 USD
102891-270 102895-310
tert-Butyl-2-phenyl-3-thioxo-1,4,8-triazaspiro[4.5]dec-1-ene-8-carboxylate ≥97%
tert-Butyl-2-phenyl-3-thioxo-1,4,8-triazaspiro[4.5]dec-1-ene-8-carboxylate

Hazard: Irritant

Formula: C₁₈H₂₃N₃O₂S
MW: 345.47 g/mol
MDL Number: MFCD14726759
CAS Number: 892289-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR