1-(3-Cyanoquinolin-4-yl)piperidine-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

115047-500MG 115047-1G
102895-282EA 693.85 USD
102895-282 102895-284
1-(3-Cyanoquinolin-4-yl)piperidine-4-carboxylic acid ≥95%
1-(3-Cyanoquinolin-4-yl)piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₅N₃O₂
MDL Number: MFCD13762738
CAS Number: 1156272-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR