8-Methyl-3-phenyl-1,4,8-triazaspiro[4.5]dec-3-ene-2-thione ≥97%

Supplier: Matrix Scientific

115043-500MG 115043-1G
102895-266EA 693.85 USD
102895-266 102895-268
8-Methyl-3-phenyl-1,4,8-triazaspiro[4.5]dec-3-ene-2-thione ≥97%
8-Methyl-3-phenyl-1,4,8-triazaspiro[4.5]dec-3-ene-2-thione

Hazard: Irritant

Formula: C₁₄H₁₇N₃S
MW: 259.38 g/mol
MDL Number: MFCD14726872
CAS Number: 892299-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR