3-Iodobenzoyl chloride 97

Supplier: Matrix Scientific

Synonyms: m-Iodobenzoyl chloride

114302-1G 114302-10G
102893-772EA 96.92 USD
102893-772 102898-386
3-Iodobenzoyl chloride 97
3-Iodobenzoyl chloride

Hazard: Irritant

Formula: C₇H₄ClIO
MW: 266.47 g/mol
MDL Number: MFCD02093934
CAS Number: 1711-10-0
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR