4-Chlorophenylhydrazine ≥97%

Supplier: Matrix Scientific

Synonyms: (4-Chlorophenyl)hydrazine

114164-1G 114164-10G
102893-480EA 109.23 USD
102893-480 102893-482
4-Chlorophenylhydrazine ≥97%
4-Chlorophenylhydrazine

Hazard: Irritant

Formula: C₆H₇ClN₂
MW: 142.59 g/mol
MDL Number: MFCD00038132
CAS Number: 1073-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR