N-Carbobenzoxy-L-serine methyl ester ≥97%

Supplier: Matrix Scientific

Synonyms: Z-Ser-OMe, CBZ-L-Serine methyl ester, N-Cbz-L-serine Methyl Ester, Methyl ((benzyloxy)carbonyl)-L-serinate

113985-1G 113985-10G
102898-346EA 60 USD
102898-346 102898-348
N-Carbobenzoxy-L-serine methyl ester ≥97%
N-Carbobenzoxy-L-serine methyl ester

Hazard: Irritant

Formula: C₁₂H₁₅NO₅
MW: 253.25 g/mol
MDL Number: MFCD00077039
CAS Number: 1676-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR