Diethyl ethoxymethylenemalonate ≥97%
Supplier: Matrix Scientific
Synonyms:
Diethyl 2-(ethoxymethylene)malonate
113980-10G
113980-50G
102893-076EA
41.54
USD
102893-076
102893-078
Diethyl ethoxymethylenemalonate ≥97%
Diethyl ethoxymethylenemalonate
Hazard: Irritant
Formula:
C₁₀H₁₆O₅ MW: 216.23 g/mol |
MDL Number:
MFCD00009148 CAS Number: 87-13-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...