Methyl-2-amino-4-methylthiophene-3-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 2-amino-4-methylthiophene-3-carboxylate

113974-1G 113974-10G
102893-060EA 60 USD
102893-060 102893-062
Methyl-2-amino-4-methylthiophene-3-carboxylate ≥97%
Methyl-2-amino-4-methylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₇H₉NO₂S
MW: 171.22 g/mol
MDL Number: MFCD01070843
CAS Number: 4651-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR