3-Amino-1H-1,2,4-triazole ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Amino-1,2,4-triazole, Amitrole, 3-Amino-1H-1,2,4-triazole

113538-1G 113538-50G
102892-386EA 15.38 USD
102892-386 102892-390
3-Amino-1H-1,2,4-triazole ≥97%
3-Amino-1H-1,2,4-triazole

Hazard: Irritant-Harmful/Refrigerate/Stored under Argon

Formula: C₂H₄N₄
MW: 84.08 g/mol
Melting Pt: 147…153 °C
MDL Number: MFCD00053362
CAS Number: 61-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR