4-Bromo-2-pyridinecarboxamide

Supplier: Matrix Scientific

Synonyms: 4-Bromopicolinamide

113442-1G 113442-5G
102892-164EA 290.77 USD
102892-164 102892-166
4-Bromo-2-pyridinecarboxamide
4-Bromo-2-pyridinecarboxamide

Hazard: Irritant

Formula: C₆H₅BrN₂O
MW: 201.03 g/mol
MDL Number: MFCD04113813
CAS Number: 62150-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR