4-Chloro-3-methoxy-2-methylpyridine

Supplier: Matrix Scientific

113316-1G 113316-5G
102897-740EA 684.62 USD
102897-740 102897-742
4-Chloro-3-methoxy-2-methylpyridine
4-Chloro-3-methoxy-2-methylpyridine

Hazard: Irritant

Formula: C₇H₈ClNO
MDL Number: MFCD16610605
CAS Number: 107512-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR