6-Chloro-3-oxo-2,3-dihydropyridazine-4-carboxylic acid methyl ester

Supplier: Matrix Scientific

113160-1G 113160-5G
102897-760EA 727.69 USD
102897-760 102897-762
6-Chloro-3-oxo-2,3-dihydropyridazine-4-carboxylic acid methyl ester
6-Chloro-3-oxo-2,3-dihydropyridazine-4-carboxylic acid methyl ester

Hazard: Irritant

Formula: C₆H₅ClN₂O₃
MDL Number: MFCD16250585
CAS Number: 89581-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR