Diisopropyl-3,3-difluorocyclobutane-1,1-dicarboxylate

Supplier: Matrix Scientific

Synonyms: 1,1-bis(propan-2-yl) 3,3-difluorocyclobutane-1,1-dicarboxylate

113051-1G 113051-5G
102897-590EA 727.69 USD
102897-590 102897-592
Diisopropyl-3,3-difluorocyclobutane-1,1-dicarboxylate
Diisopropyl-3,3-difluorocyclobutane-1,1-dicarboxylate

Hazard: Irritant

Formula: C₁₂H₁₈F₂O₄
MW: 264.27 g/mol
MDL Number: MFCD23105827
CAS Number: 1225532-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR