1-Methyl-3-phenyl-1H-pyrazole-5-carbaldehyde ≥95%

Supplier: Matrix Scientific

115793-1G 115793-5G
102897-652EA 633.85 USD
102897-652 102897-654
1-Methyl-3-phenyl-1H-pyrazole-5-carbaldehyde ≥95%
1-Methyl-3-phenyl-1H-pyrazole-5-carbaldehyde

Hazard: Irritant

Formula: C₁₁H₁₀N₂O
MDL Number: MFCD08271941
CAS Number: 864068-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR