5-Chloro-2-methyl-3-nitropyridine

Supplier: Matrix Scientific

113402-1G 113402-5G
102892-078EA 150.77 USD
102892-078 102892-080
5-Chloro-2-methyl-3-nitropyridine
5-Chloro-2-methyl-3-nitropyridine

Hazard: Irritant

Formula: C₆H₅ClN₂O₂
MW: 172.57 g/mol
MDL Number: MFCD18255264
CAS Number: 1211533-93-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR