4-(Trifluoromethoxy)hydrocinnamic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 3-[4-(Trifluoromethoxy)phenyl]propanoic acid, 3-(4-(Trifluoromethoxy)phenyl)propanoic acid

113630-1G 113630-10G
102897-940EA 86.15 USD
102897-940 102897-942
4-(Trifluoromethoxy)hydrocinnamic acid ≥97%
4-(Trifluoromethoxy)hydrocinnamic acid

Hazard: Corrosive

Formula: C₁₀H₉F₃O₃
MW: 234.18 g/mol
MDL Number: MFCD01076409
CAS Number: 886499-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR