5-(1,3-Benzodioxol-5-yl)cyclohexane-1,3-dione ≥97%

Supplier: Matrix Scientific

114892-1G 114892-10G
102894-918EA 303.08 USD
102894-918 102894-920
5-(1,3-Benzodioxol-5-yl)cyclohexane-1,3-dione ≥97%
5-(1,3-Benzodioxol-5-yl)cyclohexane-1,3-dione

Hazard: Irritant

Formula: C₁₃H₁₂O₄
MDL Number: MFCD00111711
CAS Number: 55579-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR