6-Iodo-2-oxaspiro[3.3]heptane ≥95%

Supplier: Matrix Scientific

115901-250MG 115901-500MG 115901-1G
102898-700EA 610.77 USD
102898-700 102898-702 102898-704
6-Iodo-2-oxaspiro[3.3]heptane ≥95%
6-Iodo-2-oxaspiro[3.3]heptane

Hazard: Irritant

Formula: C₆H₉IO
MW: 224.04 g/mol
MDL Number: MFCD22419348
CAS Number: 1363381-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR