3-[(Phenylmethyl)amino]-3-oxetanemethanol ≥95%

Supplier: Matrix Scientific

115603-500MG 115603-1G
102896-992EA 2416.92 USD
102896-992 102896-994
3-[(Phenylmethyl)amino]-3-oxetanemethanol ≥95%
3-[(Phenylmethyl)amino]-3-oxetanemethanol

Hazard: Irritant

Formula: C₁₁H₁₅NO₂
MDL Number: MFCD18762004
CAS Number: 1195684-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR