exo-8-Boc-3-cyano-8-azabicyclo[3.2.1]octane

Supplier: Matrix Scientific

Synonyms: exo-tert-Butyl 3-cyano-8-azabicyclo[3.2.1]octane-8-carboxylate

113203-1G 113203-5G
102891-654EA 727.69 USD
102891-654 102891-656
exo-8-Boc-3-cyano-8-azabicyclo[3.2.1]octane
exo-8-Boc-3-cyano-8-azabicyclo[3.2.1]octane

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₂
MW: 236.32 g/mol
MDL Number: MFCD23381617
CAS Number: 1147558-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR